Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000378.4 | NP_000369.3 | 9 | 2773 | |
NM_001198551.1 | NP_001185480.1 | 10 | 2276 | ||
NM_001198552.1 | NP_001185481.1 | 9 | 2234 | ||
NM_024424.3 | NP_077742.2 | 10 | 2824 | ||
NM_024426.4 | NP_077744.3 | 10 | 2833 | ||
GenBank mRNA | AB971668.1 | - | 10 | 2992 | |
AK093168.1 | - | 9 | 2230 | ||
BC032861.2 | - | 10 | 2266 | 4827849 | |
BC046461.1 | - | 10 | 2833 | ||
M30393.1 | - | 9 | 2123 | ||
X51630.1 | - | 10 | 2813 |