Gene Transcripts

Gene Symbol
SLC25A32
Entrez Gene ID
81034
Gene Name
solute carrier family 25 member 32
Gene Aliases
MFT, MFTC, RREI
Chromosome Location
Chr. 8: 103398638 - 103415335 on Build GRCh38
UniGene ID
Hs.532265
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030780.4 NP_110407.2 5 1042
NR_102337.1 4 956
NR_102338.1 6 1151
XM_017013877.1 XP_016869366.1 5 945
GenBank mRNA AF283645.1 - 5 772
AK027531.1 - 5 811
AK027787.1 - 5 776
AK125492.1 - 6 760
AK300748.1 - 4 690
BC021893.1 - 5 750 4524907
DC331449.1 - 4 454

View Full Product Details