Gene Transcripts
- Gene Symbol
- TBC1D23
- Entrez Gene ID
- 55773
- Gene Name
- TBC1 domain family member 23
- Gene Aliases
- NS4ATP1
- Chromosome Location
- Chr. 3: 100260817 - 100325252 on Build GRCh38
- UniGene ID
- Hs.477003
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199198.2 |
NP_001186127.1
|
4, 5 |
686 |
|
NM_018309.4 |
NP_060779.2
|
4, 5 |
686 |
|
XM_011512974.2 |
XP_011511276.1
|
3, 4 |
431 |
|
XM_017006840.1 |
XP_016862329.1
|
4, 5 |
518 |
|
XM_017006841.1 |
XP_016862330.1
|
4, 5 |
503 |
|
GenBank mRNA |
AB449910.1 |
- |
4, 5 |
484 |
|
AK001750.1 |
- |
1, 2 |
172 |
|
AK001908.1 |
- |
4, 5 |
491 |
|
BC020955.1 |
- |
4, 5 |
480 |
3847561 |
BX648032.1 |
- |
4, 5 |
478 |
|
View Full Product Details