Gene Transcripts

Gene Symbol
NCLN
Entrez Gene ID
56926
Gene Name
nicalin
Gene Aliases
NET59
Chromosome Location
Chr. 19: 3185877 - 3209575 on Build GRCh38
UniGene ID
Hs.657032
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321463.1 NP_001308392.1 15 2161
NM_020170.3 NP_064555.2 15 2164
GenBank mRNA AL136567.1 - 15 1995
AL365369.1 - 3 507
BC003076.2 - 7 851
BC007275.1 - 9 1140
BC025926.1 - 15 2073 3509966

View Full Product Details