Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001205262.2 | NP_001192191.1 | 4 | 504 | |
GenBank mRNA | BC033710.2 | - | 3 | 389 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256141.1 | NP_001243070.1 | 2 | 527 | |
GenBank mRNA | AF007866.1 | - | 2 | 479 |