Gene Transcripts

Gene Symbol
CDCA7L
Entrez Gene ID
55536
Gene Name
cell division cycle associated 7 like
Gene Aliases
JPO2, R1, RAM2
Chromosome Location
Chr. 7: 21900899 - 21945924 on Build GRCh38
UniGene ID
Hs.744069
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127370.2 NP_001120842.1 4, 5 473
NM_018719.4 NP_061189.2 3, 4 419
GenBank mRNA AK022955.1 - 3, 4 394
AK095965.1 - 4, 5 427
AK292490.1 - 3, 4 394
AY161168.1 - 3, 4 296
AY161169.1 - 3, 4 299
BC009352.2 - 3, 4 311 4139111
BC014630.2 - 2, 3 263
BC025242.1 - 3, 4 373 4842169
BC032576.1 - 3, 4 384 5491780
BC039823.1 - 3, 4 383 6057601
DA041070.1 - 3, 4 419

View Full Product Details