Gene Transcripts

Gene Symbol
ZDHHC9
Entrez Gene ID
51114
Gene Name
zinc finger DHHC-type containing 9
Gene Aliases
CGI89, CXorf11, DHHC9, MMSA1, MRXSZ, ZDHHC10, ZNF379, ZNF380
Chromosome Location
Chr. X: 129803288 - 129843934 on Build GRCh38
UniGene ID
Hs.193566
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008222.2 NP_001008223.1 10 1648
NM_016032.3 NP_057116.2 11 1628
GenBank mRNA AB209807.1 - 10 1567
AF151847.1 - 11 1516
AK001112.1 - 11 1578
AK001424.1 - 8 962
AK001524.1 - 10 1379
AK096967.1 - 11 1279
AL161962.1 - 11 1505
AY358558.1 - 11 1560
AY952881.8 - 11 1600
BC000035.2 - 10 1522 3506601
BC003128.1 - 11 1566 2957911
BC006200.2 - 11 1562 2964425
BC012826.2 - 11 1562 2964425

View Full Product Details