Gene Transcripts

Gene Symbol
STAG2
Entrez Gene ID
10735
Gene Name
stromal antigen 2
Gene Aliases
SA-2, SA2, SCC3B, bA517O1.1
Chromosome Location
Chr. X: 123960560 - 124102656 on Build GRCh38
UniGene ID
Hs.496710
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042749.2 NP_001036214.1 5 681
NM_001042750.1 NP_001036215.1 5 525
NM_001042751.1 NP_001036216.1 5 525
NM_001282418.1 NP_001269347.1 5 504
NM_006603.4 NP_006594.3 4 460
XM_005262357.2 XP_005262414.1 4 550
XM_005262358.2 XP_005262415.1 5 487
XM_005262359.3 XP_005262416.1 4 440
XM_005262360.2 XP_005262417.1 4 369
XM_005262361.2 XP_005262418.1 4 497
XM_006724727.1 XP_006724790.1 6 629
XM_006724728.3 XP_006724791.1 5 563
XM_011531253.2 XP_011529555.1 4 422
XM_017029232.1 XP_016884721.1 5 487
XM_017029233.1 XP_016884722.1 4 369
XM_017029234.1 XP_016884723.1 4 422
XM_017029235.1 XP_016884724.1 5 568
GenBank mRNA AK098737.1 - 4 504
AK313559.1 - 4 352
AL831939.2 - 4 429
BC001765.1 - 4 409 3352347
BX640970.1 - 5 536
BX641002.1 - 4 429
BX641003.1 - 5 494
CR627425.1 - 5 503
CR749433.1 - 5 494
DA214149.1 - 4 460

View Full Product Details