Gene Transcripts

Gene Symbol
MYH13
Entrez Gene ID
8735
Gene Name
myosin heavy chain 13
Gene Aliases
MyHC-IIL, MyHC-eo
Chromosome Location
Chr. 17: 10300866 - 10373005 on Build GRCh38
UniGene ID
Hs.711142
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003802.2 NP_003793.2 23 2984
GenBank mRNA AF111782.2 - 23 2984
AK294316.1 - 13 1786

View Full Product Details