Gene Transcripts
- Gene Symbol
- TCTA
- Entrez Gene ID
- 6988
- Gene Name
- T-cell leukemia translocation altered
- Gene Aliases
- Chromosome Location
- Chr. 3: 49412206 - 49416476 on Build GRCh38
- UniGene ID
- Hs.517962
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022171.2 |
NP_071503.1
|
3 |
2127 |
|
GenBank mRNA |
L41143.1 |
- |
3 |
2127 |
|
View Full Product Details