Gene Transcripts

Gene Symbol
SPATA20
Entrez Gene ID
64847
Gene Name
spermatogenesis associated 20
Gene Aliases
HEL-S-98, SSP411, Tisp78
Chromosome Location
Chr. 17: 50547089 - 50555852 on Build GRCh38
UniGene ID
Hs.103147
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258372.1 NP_001245301.1 6 853
NM_001258373.1 NP_001245302.1 7 956
NM_022827.3 NP_073738.2 7 901
GenBank mRNA AK025000.1 - 6 750
AK025622.1 - 2 142
AK092731.1 - 7 886
AK125807.1 - 9 1631
AK225189.1 - 6 750
AK291805.1 - 7 844
AK302850.1 - 5 878
AK310416.1 - 6 745
BC025255.1 - 7 789 5019010
BC065526.1 - 7 843 6149633
BC111029.1 - 6 758 5245538
CR936769.1 - 6 771
DQ238597.1 - 6 819
GQ472220.1 - 6 758
KJ806761.1 - 2 226

View Full Product Details