Gene Transcripts

Gene Symbol
Selplg
Entrez Gene ID
20345
Gene Name
selectin, platelet (p-selectin) ligand
Gene Aliases
CD162, Psgl-1, Psgl1, Selp1, Selpl
Chromosome Location
Chr. 5: 113817798 - 113832638 on Build GRCm38
UniGene ID
Mm.332590
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009151.3 NP_033177.3 2 1775
GenBank mRNA AK089214.1 - 3 1364
AK155762.1 - 2 1711
AK156933.1 - 2 1706
AK158134.1 - 2 1707
AK171693.1 - 3 1510
BC003874.1 - 3 1647 3495181
BY553294.1 - 1 355
X91144.1 - 2 1566

View Full Product Details