Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009167.3 | NP_033193.2 | 9 | 779 | |
GenBank mRNA | BC103611.1 | - | 9 | 779 | |
BC103612.1 | - | 9 | 779 | 40046306 | |
BC105644.1 | - | 9 | 779 | 40046301 | |
BC105645.1 | - | 9 | 779 | 40046303 | |
U46854.1 | - | 9 | 779 |