Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009200.3 | NP_033226.1 | 10 | 1695 | |
GenBank mRNA | AK159197.1 | - | 10 | 1696 | |
BC138120.1 | - | 10 | 1597 | 8861139 | |
BC138140.1 | - | 10 | 1597 | 8861159 | |
D83262.1 | - | 10 | 1689 |