Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001301671.1 | NP_001288600.1 | 4 | 1643 | |
NM_001301672.1 | NP_001288601.1 | 4 | 1475 | ||
NM_001301673.1 | NP_001288602.1 | 4 | 1526 | ||
NM_019929.4 | NP_064313.1 | 4 | 1517 | ||
GenBank mRNA | AF063847.1 | - | 4 | 1344 | |
AK148306.1 | - | 4 | 1381 | ||
AK150063.1 | - | 4 | 1390 | ||
AK150200.1 | - | 4 | 1390 | ||
AK162035.1 | - | 4 | 1390 | ||
AK167207.1 | - | 4 | 1358 | ||
AK168988.1 | - | 4 | 1381 | ||
AK170896.1 | - | 4 | 1575 | ||
AK171169.1 | - | 4 | 1575 |