Gene Transcripts
- Gene Symbol
- Ldha
- Entrez Gene ID
- 16828
- Gene Name
- lactate dehydrogenase A
- Gene Aliases
- Ldh1, Ldhm, l7R2
- Chromosome Location
- Chr. 7: 46845804 - 46855627 on Build GRCm38
- UniGene ID
- Mm.29324
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001136069.2 |
NP_001129541.2
|
2, 3 |
465 |
|
NM_010699.2 |
NP_034829.1
|
2, 3 |
272 |
|
GenBank mRNA |
AK089929.1 |
- |
2, 3 |
236 |
|
AK146121.1 |
- |
2, 3 |
233 |
|
AK149919.1 |
- |
2, 3 |
233 |
|
AK150156.1 |
- |
2, 3 |
236 |
|
AK167789.1 |
- |
2, 3 |
234 |
|
AK167944.1 |
- |
2, 3 |
234 |
|
AK168346.1 |
- |
2, 3 |
233 |
|
AK168920.1 |
- |
2, 3 |
238 |
|
AK170386.1 |
- |
2, 3 |
237 |
|
AK170708.1 |
- |
2, 3 |
234 |
|
AK171373.1 |
- |
2, 3 |
235 |
|
AK214480.1 |
- |
1, 2 |
157 |
|
AU079257.1 |
- |
|
273 |
|
BC005509.1 |
- |
1, 2 |
84 |
|
BC066858.1 |
- |
2, 3 |
219 |
|
BC094019.1 |
- |
2, 3 |
189 |
4195220 |
BC094428.1 |
- |
2, 3 |
226 |
30700835 |
BY227595.1 |
- |
2, 3 |
272 |
|
U13687.1 |
- |
2, 3 |
332 |
|
View Full Product Details