Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001080388.2 | NP_001073857.1 | 12 | 1755 | |
NM_001080389.2 | NP_001073858.1 | 12 | 1755 | ||
NM_001080390.2 | NP_001073859.1 | 12 | 1472 | ||
NM_007928.3 | NP_031954.2 | 12 | 1755 | ||
GenBank mRNA | AK045329.1 | - | 12 | 1329 | |
AK154864.1 | - | 12 | 1472 | ||
AK172444.1 | - | 12 | 1715 | ||
AK172664.1 | - | 12 | 1398 | ||
AK211690.1 | - | 3 | 203 | ||
AK220191.1 | - | 13 | 1321 | ||
BC058556.1 | - | 12 | 1692 | 5289799 | |
CX210531.1 | - | 2 | 118 | ||
X70764.1 | - | 12 | 1203 |