Gene Transcripts

Gene Symbol
REEP5
Entrez Gene ID
7905
Gene Name
receptor accessory protein 5
Gene Aliases
C5orf18, D5S346, DP1, TB2, YOP1, Yip2e
Chromosome Location
Chr. 5: 112876384 - 112922334 on Build GRCh38
UniGene ID
Hs.429608
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005669.4 NP_005660.4 4 623
XM_017009844.1 XP_016865333.1 4 540
GenBank mRNA AK294496.1 - 3 609
AK295436.1 - 3 375
AK299902.1 - 5 887
AK300048.1 - 4 441
AK310729.1 - 4 987
AK312522.1 - 4 528
AY562243.1 - 4 479
BC000232.1 - 4 504 3350749
BC065926.1 - 4 482 6164621
BX648005.1 - 4 604
M74090.1 - 4 505

View Full Product Details