Gene Transcripts

Gene Symbol
Tdpoz5
Entrez Gene ID
399676
Gene Name
TD and POZ domain containing 5
Gene Aliases
Chromosome Location
Chr. 3: 93960292 - 93961314 on Build GRCm38
UniGene ID
Mm.454051
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207273.2 NP_997156.2 457
GenBank mRNA BC111104.1 - 1 455

Gene Symbol
Gm10697
Entrez Gene ID
100042761
Gene Name
predicted gene 10697
Gene Aliases
ENSMUSG00000074425
Chromosome Location
Chr. 3: 94071622 - 94072644 on Build GRCm38
UniGene ID
Mm.484910
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163731.1 NP_001157203.1 1 457

View Full Product Details