Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_198609.2 | NP_941011.1 | 4 | 354 | |
GenBank mRNA | AK153492.1 | - | 4 | 366 | |
AK160194.1 | - | 4 | 366 | ||
AK164856.1 | - | 4 | 351 | ||
BC003885.1 | - | 4 | 354 | 3590401 | |
BC089481.1 | - | 4 | 351 | 30254073 |