Gene Transcripts

Gene Symbol
Lao1
Entrez Gene ID
100470
Gene Name
L-amino acid oxidase 1
Gene Aliases
AW990848
Chromosome Location
Chr. 4: 118961967 - 118968910 on Build GRCm38
UniGene ID
Mm.26698
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133892.4 NP_598653.3 7 931
GenBank mRNA AB034801.2 - 7 931
AK052781.1 - 7 902
BC017599.1 - 7 1320 4487137
BC023198.1 - 6 777

View Full Product Details