Gene Transcripts

Gene Symbol
Tcta
Entrez Gene ID
102791
Gene Name
T cell leukemia translocation altered gene
Gene Aliases
9130410M22Rik, AW553637, C85065, Tctal
Chromosome Location
Chr. 9: 108302958 - 108306125 on Build GRCm38
UniGene ID
Mm.24670
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133986.1 NP_598747.1 3 371
GenBank mRNA AK133663.1 - 410
BC019397.1 - 3 371 3672301

View Full Product Details