Gene Transcripts
- Gene Symbol
- Slc25a33
- Entrez Gene ID
- 70556
- Gene Name
- solute carrier family 25, member 33
- Gene Aliases
- 5730438N18Rik, Pnc1
- Chromosome Location
- Chr. 4: 149744036 - 149774267 on Build GRCm38
- UniGene ID
- Mm.41877
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK017628.1 |
- |
2 |
225 |
|
View Full Product Details