Gene Transcripts

Gene Symbol
TIMM17A
Entrez Gene ID
10440
Gene Name
translocase of inner mitochondrial membrane 17 homolog A (yeast)
Gene Aliases
TIM17, TIM17A
Chromosome Location
Chr. 1: 201955491 - 201970661 on Build GRCh38
UniGene ID
Hs.20716
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006335.2 NP_006326.1 4 255
GenBank mRNA AF106622.1 - 4 226
AK023063.1 - 1 1429
AK315602.1 - 4 243
AU123184.1 - 4 255
BC000294.2 - 4 225 2819455
BC004439.1 - 4 225 2819455
BC007106.1 - 4 236 4284092
BC009784.1 - 4 247 3683125
BC013823.1 - 4 214
BC015098.1 - 4 253 3686369
BC016817.1 - 2 110
BC020833.1 - 4 233 4773149
X97544.1 - 4 232

View Full Product Details