Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001162500.1 | NP_001155972.1 | 13 | 1508 | |
NM_022321.4 | NP_071716.3 | 13 | 1463 | ||
GenBank mRNA | AK028329.1 | - | 13 | 1464 | |
AK037365.1 | - | 13 | 1436 | ||
AK040914.1 | - | 13 | 1394 | ||
AK079881.1 | - | 13 | 1389 | ||
AK088419.1 | - | 13 | 1413 | ||
BC011200.1 | - | 13 | 1417 | 4190510 |