Gene Transcripts
- Gene Symbol
- TUBGCP2
- Entrez Gene ID
- 10844
- Gene Name
- tubulin gamma complex associated protein 2
- Gene Aliases
- GCP-2, GCP2, Grip103, SPBC97, Spc97p, h103p, hGCP2, hSpc97
- Chromosome Location
- Chr. 10: 133278630 - 133312337 on Build GRCh38
- UniGene ID
- Hs.523370
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001256617.1 |
NP_001243546.1
|
10, 11 |
1825 |
|
NM_001256618.1 |
NP_001243547.1
|
8, 9 |
1552 |
|
NM_006659.3 |
NP_006650.1
|
9, 10 |
1741 |
|
NR_046330.1 |
|
9, 10 |
2125 |
|
XM_017015595.1 |
XP_016871084.1
|
2, 3 |
210 |
|
XM_017015596.1 |
XP_016871085.1
|
2, 3 |
210 |
|
GenBank mRNA |
AF042379.1 |
- |
9, 10 |
1431 |
|
AK092113.1 |
- |
9, 10 |
2125 |
|
AK096087.1 |
- |
1, 2 |
1219 |
|
AK223586.1 |
- |
9, 10 |
1632 |
|
AK297251.1 |
- |
8, 9 |
1274 |
|
AK299564.1 |
- |
9, 10 |
1295 |
|
AK314520.1 |
- |
9, 10 |
1463 |
|
BC093770.1 |
- |
8, 9 |
1420 |
7939615 |
BC111957.1 |
- |
8, 9 |
1420 |
8327425 |
BC143247.1 |
- |
9, 10 |
1504 |
9051749 |
View Full Product Details