Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019723.2 | NP_062697.1 | 10 | 1826 | |
GenBank mRNA | AB018436.1 | - | 10 | 1826 | |
AK080177.1 | - | 9 | 2029 | ||
AK133431.1 | - | 10 | 1755 | ||
AK161508.1 | - | 10 | 1722 | ||
BC132147.1 | - | 10 | 1789 | 40130424 | |
BC137908.1 | - | 10 | 1789 | 8860927 |