Gene Transcripts

Gene Symbol
Tpmt
Entrez Gene ID
22017
Gene Name
thiopurine methyltransferase
Gene Aliases
AW106912
Chromosome Location
Chr. 13: 47021987 - 47043338 on Build GRCm38
UniGene ID
Mm.10169
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016785.2 NP_058065.2 2 267
GenBank mRNA AF037043.1 - 2 176
AF037044.1 - 1 127
AF037045.1 - 1 127
AF046887.1 - 2 253
AK002335.1 - 2 174
AK019205.1 - 2 172
BC021598.1 - 1 154 5098857
CF106558.1 - 2 189

View Full Product Details