Gene Transcripts

Gene Symbol
Tfcp2
Entrez Gene ID
21422
Gene Name
transcription factor CP2
Gene Aliases
CP-2, CP2, D230015P20Rik, LBP-1c, LBP-1d, LBP1, LSF, Tcfcp2, UBP-1
Chromosome Location
Chr. 15: 100502748 - 100552014 on Build GRCm38
UniGene ID
Mm.219040
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289603.1 NP_001276532.1 3 328
NM_033476.3 NP_258437.1 3 328
GenBank mRNA AK051891.1 - 2 327
AK131989.1 - 2 527
AK146503.1 - 3 329
AK147898.1 - 2 537
AK158682.1 - 3 248
BC022131.1 - 3 301 5098336
M84809.1 - 3 267

View Full Product Details