Gene Transcripts

Gene Symbol
NPM3
Entrez Gene ID
10360
Gene Name
nucleophosmin/nucleoplasmin 3
Gene Aliases
PORMIN, TMEM123
Chromosome Location
Chr. 10: 101781325 - 101783413 on Build GRCh38
UniGene ID
Hs.90691
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006993.2 NP_008924.1 5 493
GenBank mRNA AF081280.1 - 5 481
AY049737.1 - 5 485
BC041067.1 - 5 474 5473410
BC054868.1 - 5 479 6496849
BM739536.1 - 5 493
BM765409.1 - 5 493

View Full Product Details