Gene Transcripts

Gene Symbol
PAGE4
Entrez Gene ID
9506
Gene Name
PAGE family member 4
Gene Aliases
CT16.7, GAGE-9, GAGEC1, JM-27, JM27, PAGE-1, PAGE-4
Chromosome Location
Chr. X: 49829260 - 49834264 on Build GRCh38
UniGene ID
Hs.441038
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318877.1 NP_001305806.1 4 267
NM_007003.4 NP_008934.1 4 279
GenBank mRNA AF275258.1 - 4 265
AJ005894.1 - 4 263
AK312039.1 - 4 276
AL548759.3 - 3 230
BC010897.2 - 3 229 4043535
BG435968.1 - 4 280
BG622311.1 - 281
CR456823.1 - 3 200
CR542183.1 - 3 200
DA841276.1 - 4 261

View Full Product Details