Gene Transcripts

Gene Symbol
Prl7d1
Entrez Gene ID
84377
Gene Name
prolactin family 7, subfamily d, member 1
Gene Aliases
PRP, Plfr
Chromosome Location
Chr. 17: 38993183 - 38999651 on Build Rnor_6.0
UniGene ID
Rn.15366
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053364.1 NP_445816.1 6 739
GenBank mRNA AF139809.1 - 6 739
AF226609.1 - 6 711
FQ219949.1 - 6 735
FQ220120.1 - 6 686

View Full Product Details