Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_053669.1 | NP_446121.1 | 4 | 1121 | |
XM_006249092.2 | XP_006249154.1 | 4 | 1873 | ||
XM_006249093.2 | XP_006249155.1 | 4 | 1874 | ||
XM_008769112.1 | XP_008767334.1 | 5 | 1672 | ||
XM_008769113.1 | XP_008767335.1 | 4 | 1136 | ||
GenBank mRNA | AF095576.1 | - | 4 | 1121 |