Gene Transcripts

Gene Symbol
Mfng
Entrez Gene ID
315119
Gene Name
MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Gene Aliases
Chromosome Location
Chr. 7: 119978983 - 119996824 on Build Rnor_6.0
UniGene ID
Rn.102109
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199110.1 NP_954541.1 8 1549
GenBank mRNA BC061801.1 - 8 1549

View Full Product Details