Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012430.4 | NP_036562.2 | 2 | 212 | |
XM_011512673.2 | XP_011510975.1 | 2 | 1728 | ||
XM_011512675.2 | XP_011510977.1 | 3 | 379 | ||
XM_011512676.2 | XP_011510978.1 | 2 | 268 | ||
GenBank mRNA | AF100749.1 | - | 2 | 194 | |
AK057587.1 | - | 1 | 1028 | ||
AK315771.1 | - | 2 | 181 | ||
BC007122.1 | - | 1 | 160 | 3459649 | |
BG613364.1 | - | 2 | 213 | ||
DA308910.1 | - | 2 | 212 |