Gene Transcripts

Gene Symbol
YIF1B
Entrez Gene ID
90522
Gene Name
Yip1 interacting factor homolog B, membrane trafficking protein
Gene Aliases
FinGER8
Chromosome Location
Chr. 19: 38303560 - 38315976 on Build GRCh38
UniGene ID
Hs.280741
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039671.2 NP_001034760.1 3 359
NM_001039672.2 NP_001034761.1 3 377
NM_001039673.2 NP_001034762.1 3 368
NM_001145461.1 NP_001138933.1 3 353
NM_001145462.1 NP_001138934.1 3 329
NM_001145463.1 NP_001138935.1 3 368
XM_005259385.3 XP_005259442.1 3 590
XM_017027449.1 XP_016882938.1 3 378
XM_017027450.1 XP_016882939.1 3 376
XM_017027451.1 XP_016882940.1 3 374
GenBank mRNA AF258578.3 - 2 551
AK304479.1 - 2 420
AL833382.1 - 3 325
AY358965.1 - 2 496
BC007644.1 - 3 316
BC014974.1 - 3 322 4865479
BC091477.1 - 3 351 30344388
BE877226.1 - 3 305
BG765536.1 - 3 310
BM703847.1 - 3 359
BX379501.2 - 3 295
CA487891.1 - 3 311
CA488303.1 - 3 306
CF127536.1 - 3 323

View Full Product Details