Gene Transcripts

Gene Symbol
SLC2A8
Entrez Gene ID
29988
Gene Name
solute carrier family 2 member 8
Gene Aliases
GLUT8, GLUTX1
Chromosome Location
Chr. 9: 127397138 - 127407898 on Build GRCh38
UniGene ID
Hs.179522
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271712.1 NP_001258641.1 7 1009
NM_014580.4 NP_055395.2 9 1379
NR_073416.1 6 812
XM_006717084.3 XP_006717147.1 9 1374
XM_011518602.2 XP_011516904.1 9 1295
XM_011518603.2 XP_011516905.1 7 1034
XM_011518604.2 XP_011516906.1 7 958
GenBank mRNA AJ245937.1 - 9 1289
BC019043.2 - 9 1331 4641145
BM044230.1 - 6 713
Y17801.1 - 9 1312

View Full Product Details