Gene Transcripts
- Gene Symbol
- TGFB1I1
- Entrez Gene ID
- 7041
- Gene Name
- transforming growth factor beta 1 induced transcript 1
- Gene Aliases
- ARA55, HIC-5, HIC5, TSC-5
- Chromosome Location
- Chr. 16: 31472155 - 31477960 on Build GRCh38
- UniGene ID
- Hs.513530
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC001507.3 |
- |
11 |
1551 |
|
BC001830.2 |
- |
11 |
1551 |
3030477 |
View Full Product Details