Gene Transcripts

Gene Symbol
LIMK1
Entrez Gene ID
3984
Gene Name
LIM domain kinase 1
Gene Aliases
LIMK, LIMK-1
Chromosome Location
Chr. 7: 74083777 - 74122525 on Build GRCh38
UniGene ID
Hs.647035
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204426.1 NP_001191355.1 8 1084
NM_002314.3 NP_002305.1 9 1306
GenBank mRNA AB209557.1 - 6 778
AF134379.1 - 9 1031
AK026544.1 - 3 342
AK290162.1 - 10 1231
AK300382.1 - 8 1023
D26309.1 - 10 1187

View Full Product Details