Gene Transcripts
- Gene Symbol
- MCCD1
- Entrez Gene ID
- 401250
- Gene Name
- mitochondrial coiled-coil domain 1
- Gene Aliases
- Chromosome Location
- Chr. 6: 31528962 - 31530231 on Build GRCh38
- UniGene ID
- Hs.558922
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC132994.1 |
- |
2 |
426 |
|
BC132996.1 |
- |
2 |
426 |
40147018 |
View Full Product Details