Gene Transcripts

Gene Symbol
Rxrb
Entrez Gene ID
361801
Gene Name
retinoid X receptor beta
Gene Aliases
RXR-beta
Chromosome Location
Chr. 20: 3822678 - 3829138 on Build Rnor_6.0
UniGene ID
Rn.49295
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206849.3 NP_996731.2 4 676
XM_006255960.2 XP_006256022.1 4 1186
XM_006255961.2 XP_006256023.1 4 679
GenBank mRNA BC099776.1 - 4 578
M81766.1 - 4 505

View Full Product Details