Gene Transcripts

Gene Symbol
Nup35
Entrez Gene ID
69482
Gene Name
nucleoporin 35
Gene Aliases
2310006I24Rik, 35kDa, 5330402E05Rik, MP44, NO44
Chromosome Location
Chr. 2: 80638812 - 80660071 on Build GRCm38
UniGene ID
Mm.29200
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190179.1 NP_001177108.1 9 1365
NM_027091.4 NP_081367.1 9 1345
GenBank mRNA AF411517.1 - 9 1324
AK089997.1 - 9 1344
AK136109.1 - 9 1309
AK145775.1 - 9 1293
BC048814.1 - 9 1300 2647417

View Full Product Details