Gene Transcripts

Gene Symbol
Xkr7
Entrez Gene ID
228787
Gene Name
X Kell blood group precursor related family member 7 homolog
Gene Aliases
AY534253
Chromosome Location
Chr. 2: 153028540 - 153060294 on Build GRCm38
UniGene ID
Mm.376111
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011732.1 NP_001011732.1 3 1325
GenBank mRNA AY534253.1 - 3 1325

View Full Product Details