Gene Transcripts

Gene Symbol
Snx7
Entrez Gene ID
310815
Gene Name
sorting nexin 7
Gene Aliases
Chromosome Location
Chr. 2: 220008463 - 220027686 on Build Rnor_6.0
UniGene ID
Rn.29845
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012083.1 NP_001012083.1 9 1495
XM_006233234.2 XP_006233296.1 9 3090
GenBank mRNA BC082033.1 - 9 1495

View Full Product Details