Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012435.2 | NP_036567.2 | 1 | 392 | |
XM_011527893.2 | XP_011526195.1 | 1 | 407 | ||
XM_011527894.2 | XP_011526196.1 | 1 | 407 | ||
XM_011527895.2 | XP_011526197.1 | 1 | 407 | ||
GenBank mRNA | AB001451.1 | - | 1 | 266 | |
AL134433.1 | - | 1 | 49 |