Gene Transcripts

Gene Symbol
AKR1C6P
Entrez Gene ID
389932
Gene Name
aldo-keto reductase family 1 member C6, pseudogene
Gene Aliases
TAKR
Chromosome Location
Chr. 10: 4871667 - 4916273 on Build GRCh38
UniGene ID
Hs.631812
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026743.1 5 506
GenBank mRNA AB037902.1 - 3 309
CB111913.1 - 5 461
DA643129.1 - 4 439

View Full Product Details