Gene Transcripts

Gene Symbol
Mmd2
Entrez Gene ID
304301
Gene Name
monocyte to macrophage differentiation-associated 2
Gene Aliases
Chromosome Location
Chr. 12: 14021724 - 14078121 on Build Rnor_6.0
UniGene ID
Rn.154557
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037217.1 NP_001032294.1 7 1302
GenBank mRNA BC161816.1 - 7 1280
CB579379.1 - 1 374

View Full Product Details