Gene Transcripts

Gene Symbol
Noc2l
Entrez Gene ID
313777
Gene Name
nucleolar complex associated 2 homolog (S. cerevisiae)
Gene Aliases
RGD1309387
Chromosome Location
Chr. 5: 173660921 - 173672699 on Build Rnor_6.0
UniGene ID
Rn.3274
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033897.1 NP_001029069.1 7 946
GenBank mRNA BC101911.1 - 7 946

View Full Product Details