Gene Transcripts

Gene Symbol
Naif1
Entrez Gene ID
71254
Gene Name
nuclear apoptosis inducing factor 1
Gene Aliases
2310007O20Rik, 4933440H19Rik
Chromosome Location
Chr. 2: 32450457 - 32455476 on Build GRCm38
UniGene ID
Mm.102341
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194335.2 NP_919316.1 2 2988
GenBank mRNA AK009216.1 - 225
AK029812.1 - 2 2989
BC057613.1 - 1244 6401522

View Full Product Details