Gene Transcripts
- Gene Symbol
- Rdh18-ps
- Entrez Gene ID
- 380674
- Gene Name
- retinol dehydrogenase 18, pseudogene
- Gene Aliases
- Rdh18
- Chromosome Location
- Chr. 10: 127824216 - 127846565 on Build GRCm38
- UniGene ID
- Mm.384871
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY053573.1 |
- |
3 |
692 |
|
View Full Product Details